Pipette tips, WHEATON®
Macro, 10 ml, +, +, Case of 100 tips
Codice catalogo WHEA851358
Nome del fornitore: DWK Life Sciences
Vai a
Product Details & Documents
These high quality tips are made of high grade, virgin, metal-free PP.
- Compatible with most macro pipettes
- Precision injection molding care for air-tight fit
- Free from trace metals for excellent accuracy and reproducibility
Specifiche
- Confezione:Case of 100 tips
- Sterili:+
- Volume:10 ml
- Seq:03
- Stile punta:Macro
- Filtro:+