Macron Fine Chemicals™ CHEM-SOLV®, Laboratory Glassware Cleaner
Katalognr. 2157-04
Anbieter: MACRON AVANTOR BRAND
Technische Daten
- Descrizione:CHEM-SOLV®, laboratory glassware cleaner
- Seq:01
- Tipo di conf.:Jerrican
- Confezione:Narrow mouth polyethylene bottle
- Conf:1 qt US
Technische Daten
Über diesen Artikel
CHEM-SOLV® is a non chromic acid cleaner suitable for removing silicone greases, dried blood, and other organic residues from laboratory glass and plasticware. It is environmentally friendly and safer alternative for chromic and sulphuric acid solution.
- Free of interfering ions
- Ideal for cleaning labware used in sensitive analytical procedures and organic analysis
- Cleans glassware without scrubbing