Sublimation apparatus
Sublimation apparatus with glass stopcock (14,5/2,5 mm) and cold finger, 29/32 NS
Catalog # NEUB2485-06-29
Supplier: NEUBERT VOLUME GLASSWAERE
Jump to:
Product Details & Documents
This sublimation apparatus, made of borosilicate glass 3.3, is for working under vacuum.Total height: 200 mm.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
- Cono femmina:29/32 NS
- Descrizione:Sublimation apparatus with glass stopcock (14,5/2,5 mm) and cold finger