Immobilised Pepstatin for purification of Cathepsins/Pepsin/Pepstatin binding molecules
Codice catalogo 786-789
Nome del fornitore: G-Biosciences
Numero CAS:
Specifiche
- Conf:5 mL
- Descrizione:Immobilised Pepstatin
Specifiche
Informazioni su questo articolo
Immobilised Pepstatin is for the purification of cathepsins, pepsin, bacterial aspartic proteases, HIV proteases and other molecules that bind pepstatin.