Needles, Atraucan®
Atraucan® Paed needles for pediatric anesthesia, 26, 25 mm, 0,47 mm, marrone
BRAU4504771
: B. Braun
Special atraumatic needles for spinal anesthesia.
- Patented two-zone atraumatic bevel
- Transparent and ergonomic needle hub design
- Colour-coded stylet hub
Specifically designed for applications in pediatrics.
- Codice colore:marrone
- Ø:0,47 mm
- Gauge:26
- Descrizione:Atraucan® Paed needles for pediatric anesthesia
- Lunghezza:25 mm