CytoScan™ Cell proliferation assay kit, WST-1
Référence 786-212
Fournisseur: G-Biosciences
Numéro CAS:
Spécifications
- Conf:500 Assays
- Descrizione:CytoScan™ WST-1 Cell Proliferation Assay Kit
- Test per kit:500 assays
Spécifications
À propos de cet article
CytoScan™ WST-1 Cell Proliferation Assay principle is based upon the reduction of the tetrazolium salt WST-1 to formazan by cellular dehydrogenases. The generation of the dark yellow coloured formazan is measured at 420 to 480 nm (optimal at 440 nm) and is directly correlated to cell number.
- Sensitive and accurate assay
- Colorimetric assay
- No washing, harvesting or solubilization required
- For the detection of cell toxicity, death, viability or proliferation
- Ideal for high throughput screening