VWR®, pH electrode, flat
pH electrode, flat, 100 psig rated, with double reference junction, for use with flow cell, 0 - 14, BNC, 0,7 m cable
Codice catalogo 662-1381
Nome del fornitore: VWR Collection
Vai a
Product Details & Documents
This flat surface electrode is a combination pH/reference electrode with double reference junction design. It includes both a pH and reference half cell. The reference electrode is a sealed, gel-filled design and includes a peripheral, semi-porous polyethylene junction.
- Designed for use with a variety of instrumental techniques such as FIA, HPLC and IEF
- Suitable for continuously monitoring the pH of low volume flows in bench scale chemical systems
Max pressure: 6894,7 mbar (100 psi).
Informazioni per l'ordine: To be used with a suitable flow cell. Please contact your local Avantor sales office for further information.
Documents
662-1381
Immagini 360
Specifiche
- Connettore:BNC, 0,7 m cable
- Descrizione:pH electrode, flat, 100 psig rated, with double reference junction, for use with flow cell
- pH:0 - 14