Software, IRsolution Agent
IR solution agent software version 2.2 (single user license)
Katalognr. SHIM206-21600-92
Anbieter: SHIMADZU
Direkt zu
Product Details & Documents
IRsolution Agent is a tool for managing data from IRsolution results in a database. This allows managing analytical data records over their entire life cycle, from creation (measurement), review, approval, storage, viewing, backup, and disposal.
- Provides security management and electronic signature functinos for data stored and recorded in a database
- Measurement data is automatically saved in a database to allow easy searching of target data at a later time
- Spectral waveform data, peak detection tables and other information stored in the database can be viewed directly
Certifications: Compliant with FDA 21 CFR Part 11 (electronic records and electronic signatures).
Technische Daten
- Descrizione:IR solution agent software version 2.2 (single user license)