Chemical resistant gloves, AlphaTec® AquaDri® 58-330/58-335
: Ansell
These two-layer gloves protect and keep hands dry. An inner soft foam coating with proprietary AquaDri® technology offers unrivalled moisture absorption: Up to 10 times more than traditional cotton-flocked linings on average. The nitrile film provides advanced chemical protection against incidental exposure to bases, oils, fuels, some solvents, greases and animal fats.
- Reversed-lozenge finish provides improved dry and wet grip, delivering optimal results in dry or wet conditions
- High performance nitrile compound offers superior snag, puncture and abrasion protection compared to rubber or neoprene
- Foldable cuff gutter prevents dripping onto the forearm
Thickness:
58-330: 0,3 mm (Nitrile) +0,35 mm (AquaDri® lining)
58-335: 0,45 mm (Nitrile) +0,35 mm (AquaDri® lining)
Thickness: 58-330: 0,3 mm (Nitrile) + 0,35 mm (AquaDri® lining) 58-335: 0,45 mm (Nitrile) + 0,35 mm (AquaDri® lining)
Colour: Turquoise or green
EN 374 (JKL), EN 388 (4101), EN 1149; Cat. III; AQL 1,5 Food contact approved
: 58-330: EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (3101X), EN 1149; Cat. III; AQL 1,5
58-335: EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (4101A), EN 1149; Cat. III; AQL 1,5
Food contact approved
58-335: EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (4101A), EN 1149; Cat. III; AQL 1,5
Food contact approved
- Catalog No:
- 112-3405
- 112-3406
- 112-3402
- 112-3403
- 112-3397
- 112-3398
- 112-3399
- 112-3400
- 112-3401
- 112-3404
- Ambidestro/mano specifico:
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- mano specifico
- Monouso/riutilizzabile:
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- riutilizzabile
- Materiale rivestimento interno:
- Fibre polimeriche
- Fibre polimeriche
- Fibre polimeriche
- Fibre polimeriche
- Fibre polimeriche
- Fibre polimeriche
- Fibre polimeriche
- Fibre polimeriche
- Fibre polimeriche
- Fibre polimeriche
- Finitura:
- Reversed lozenge
- Reversed lozenge
- Reversed lozenge
- Reversed lozenge
- Reversed lozenge
- Reversed lozenge
- Reversed lozenge
- Reversed lozenge
- Reversed lozenge
- Reversed lozenge
- Tipo polsino:
- Svasato
- Svasato
- Svasato
- Svasato
- Svasato
- Svasato
- Svasato
- Svasato
- Svasato
- Svasato
- Norma EN:
- EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (4101A)
- EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (4101A)
- EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (4101A)
- EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (4101A)
- EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (3101X)
- EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (3101X)
- EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (3101X)
- EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (3101X)
- EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (3101X)
- EN ISO 374-1:2016 Type A (JKLOPT), EN ISO 374-5:2016, EN388:2016 (4101A)
- Antistatico:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
- Approvati per il contatto con gli alimenti:
- +
- +
- +
- +
- +
- +
- +
- +
- +
- +
Frequently Bought Together