VWR®, Disposal Box for Pipette Tips
212-1833
- Descrizione:Disposal box for pipette tips, clear acrylic, with flip cover
- L×P×A:154×171×206 mm
- Conf:1
This acrylic box is ideal for the one-handed disposal of used pipette tips. The 10 mm thick acrylic provides protection from β radiation.
- Quick flip of port cover makes tip ejection easy
- Eliminates any need to handle pipette tips used with hazardous fluids
- Large hinged lid allows easy disposal of contents
W×D×H: 154×171×206 mm