Protein blocking agent, BLOT-QuickBlocker™
BLOT-QuickBlocker™ blocking agent, 175 g
Codice catalogo 786-011
Nome del fornitore: G-Biosciences
Protein blocking agent, BLOT-QuickBlocker™
Codice catalogo 786-011
Nome del fornitore: G-Biosciences
Numero CAS:
Vai a
Product Details & Documents
BLOT-QuickBlocker™ is a modified milk protein that is highly soluble and does not inhibit peroxidase detection. The modified milk protein has high blocking efficiency with a clear background.
- Readily soluble and produces semi-clear solution
- No inhibition to peroxidase
- Fat free and produces clear background
- Higher blocking efficiency
- Blocking time 30 to 60 minutes
Documents
786-011_protocol
Manuale
Specifiche
Specifiche
- Conf:175 g
- Descrizione:BLOT-QuickBlocker™ blocking agent
- Cat. No.:786-011