Optimum Growth Flask
Nome del fornitore: THOMSON INSTRUMENT
Vai a
Informazioni su questo articolo
Optimum Growth™ flasks (patented) give excellent growth and expression for both mammalian and insect cell lines.
- Baffles designed for high aeration and low shear
- Same footprint as compairable Fernbach flask
- Less foaming than disposable Fernbach flask
- Transfer cap connects directly to Wave Bags™ and bioreactors with quick connect or tube fusing
- 0,2 μm vented cap
- Individually packaged and sterilised
6 varianti
Product Details & Documents
Optimum Growth™ flasks (patented) give excellent growth and expression for both mammalian and insect cell lines.
- Baffles designed for high aeration and low shear
- Same footprint as compairable Fernbach flask
- Less foaming than disposable Fernbach flask
- Transfer cap connects directly to Wave Bags™ and bioreactors with quick connect or tube fusing
- 0,2 μm vented cap
- Individually packaged and sterilised
Documents
Certificates
Cerca per numero di lottoProduct Family Options
Informazioni sul prodotto
- CapacitàDisponibilitàPrice
- THMS931116Optimum Growth™ flask
- THMS931111Optimum Growth™ flask
- THMS931112Optimum Growth™ flask
- THMS931114Optimum Growth™ flask
- THMS931110Optimum Growth™ flask
- THMS931113Optimum Growth™ flask