Orders LinkContactUsLinkComponent
Protease Screening Kit | Avantor
Protease Screening Kit
Protease Screening Kit, 50 assays, 1 KIT
Codice catalogo 786-137
Nome del fornitore:   G-Biosciences
Protease Screening Kit
Codice catalogo 786-137
Nome del fornitore:  G-Biosciences
Numero CAS:  
Protease Screening Kit
Protease Screening Kit, 50 assays, 1 KIT
Numero CAS:  
Price
SFr. 399.00
Log in per disponibilità
1 * 1 KIT
Product Details & Documents
The Protease Screening Kit provides a simple and quick method for testing samples for proteolysis. Simply incubate sample in the reagent provided and obtain results.
The kit uses dye-labelled protein conjugate as protease substrate, which allows nanogram level detection. The absorbance of dye-labelled peptide is measured at 574 nm for determination of protease activity.
The kit is sufficient for 50 assays in a microwell format.
Dokumente
Specifiche
Specifiche
  • Conf:
    1 KIT
  • Descrizione:
    Protease Screening Kit
  • Test per kit:
    50 assays