Western blots, pre-made, mouse subcellular fraction specific blot
Catalog # TB47
Supplier: G-Biosciences
CAS Number:
Specifications
- Conf:1
- Descrizione:Mouse subcellular fraction specific blot, normal tissue
Specifications
About this item
The subcellular fractions from liver tissue are prepared by carefully dissecting out anatomically and functionally distinct regions of adult mouse livers.
Each blot contains the following mouse liver subcellular fractions: Whole liver, nuclei, mitochondria, Golgi complex, smooth endoplasmic reticulum, and rough endoplasmic reticulum.