Reference electrode, Dionex™
pH reference electrode, Ion chromatography (carbohydrate analyses/amino acid analyses)
DION044198
Marketsource
: DIONEX
Detector pH-Ag/AgCl reference electrode which can be used for carbohydrate and amino acid analysis with the Thermo Scientific™ Dionex™ ED electrochemical detector pH reference electrode.
- Per:Ion chromatography (carbohydrate analyses/amino acid analyses)
- Descrizione:pH reference electrode