Distilling bridge with Liebig condenser, Quickfit®
Codice catalogo 201-0607
Nome del fornitore: SciLabware
Specifiche
- Dimensione cono:19/26 NS
- Lunghezza mantello:155 mm
- Cono femmina:14/23 NS
- Conf:1
Specifiche
Informazioni su questo articolo
PYREX®, borosilicate glass
- Angle and position pre-defined
- Less grease contamination
- No air leakage under vacuum
- No liquid hold-up at the joints
Comprises Liebig condenser, stillhead and receiver adapter in a single piece.