Flow meters for KrosFlo® systems
Catalog # SPECACM3-FMH-01N
Anbieter: Spectrum Laboratories
Technische Daten
- Velocità del flusso:0,2 - 4,5 l/min
- Descrizione:Permeate flow meter
- Seq:01
- Conf:1
Technische Daten
Über diesen Artikel
Permeate flow meters for use with tangential flow KrosFlo® systems.