Qualitative filter papers, wet-strengthened
Supplier: PRAT DUMAS
Certificates
About this item
Grade 106: These vellum papers are designed for qualitative analysis. They are used for general laboratory filtration in universities, schools, hospital, research laboratories, food and pharmaceutical laboratories and for sweets.
Grade 609: These cotton fiber wet strengthened filter papers are used for very fast filtration of big or gelatinous precipitates.
Ordering information: For specific orders, these filters can be supplied in other diameters or in sheets.
Packaging: Filters supplied as flat in boxes of 100 filters.
Specifications
- Catalog No:
- PRATA007609
- PRATA010609
- PRATJ015609
- PRATJ020609
- PRATA015609
- PRATA025106
- PRATA013106
- PRATA007106
- PRATA009106
- PRATJ025609
- PRATA015106
- PRATA025609
- PRATJ045609
- PRATJ050609
- PRATA020609
- PRATA013609
- PRATJ040609
- PRATJ010609
- PRATJ013609
- PRATA040609
- Grado:
- 609
- 609
- 609
- 609
- 609
- 106
- 106
- 106
- 106
- 609
- 106
- 609
- 609
- 609
- 609
- 609
- 609
- 609
- 609
- 609
- Peso:
- 95 g/m²
- 95 g/m²
- 95 g/m²
- 95 g/m²
- 95 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 65 g/m²
- 95 g/m²
- 65 g/m²
- 95 g/m²
- 95 g/m²
- 95 g/m²
- 95 g/m²
- 95 g/m²
- 95 g/m²
- 95 g/m²
- 95 g/m²
- 95 g/m²
- Spessore:
- 215 μm
- 215 μm
- 215 μm
- 215 μm
- 215 μm
- 170 μm
- 170 μm
- 170 μm
- 170 μm
- 215 μm
- 170 μm
- 215 μm
- 215 μm
- 215 μm
- 215 μm
- 215 μm
- 215 μm
- 215 μm
- 215 μm
- 215 μm
- Ritenzione delle particelle:
- 20 - 25 μm
- 20 - 25 μm
- 20 - 25 μm
- 20 - 25 μm
- 20 - 25 μm
- 7 - 10 μm
- 7 - 10 μm
- 7 - 10 μm
- 7 - 10 μm
- 20 - 25 μm
- 7 - 10 μm
- 20 - 25 μm
- 20 - 25 μm
- 20 - 25 μm
- 20 - 25 μm
- 20 - 25 μm
- 20 - 25 μm
- 20 - 25 μm
- 20 - 25 μm
- 20 - 25 μm