Single stage gas regulators, BS series
Catalog # AIRL15827
Supplier: Air Liquide
Specifications
- Descrizione:Single stage gas regulator, brass, BS 50-1-2
- Seq:01
- Pressione di ingresso massima:50 bar at 15 ºC
- Pressione di uscita:0,05 - 1 bar
- Ingresso/uscita:G 3/8” female/G 3/8” female
- Tipo:BS 50-1-2
- Peso:0,5 kg
- Flusso:2,0 Nm³/h
- Conf:1
Specifications
About this item
These chromium plated brass gas regulators are used in second expansion stage in all research and industrial control laboratories, requiring extremely accurate pressure regulation. Compatible with non-corrosive pure gases (up to N60: 99,9999% - ALPHAGAZ™ 2) and mixtures.
Operating temperature: –20...+50 °C