Autosampler kits
Supplier: AGILENT HEWLETT PACKARD
Certificates
About this item
Frequently Bought Together
[EN]FILTER EXHAUST M 1 * 1 pezzi
Supplier: VWR Collection
Colonne HPLC, Chromolith®
Supplier: Merck
Avantor® ACE® AQ, HPLC Columns, 5 µm
Supplier: Avantor
Colonne SPE, J.T.Baker®, BAKERBOND spe™ C18
Supplier: Avantor
VWR®, Screw Caps for Short Thread Vials ND9
Supplier: VWR Collection
Avantor® ACE®, Guard Cartridge Holders
Supplier: Avantor
Safety funnels with lid, 'MARCO' V2.0, electrically conductive
Supplier: S.C.A.T.
Exhaust filters, V3.0
Supplier: S.C.A.T.
VWR®, Snap Cap Vials
Supplier: VWR Collection
Gel di silice 60, irregular 40 - 63 µm per cromatografia flash
Supplier: VWR Chemicals
Capillari in vetro monouso e micropipette graduate monouso
Supplier: CAMAG
Avantor® ACE® C18, colonne HPLC, 5 µm
Supplier: Avantor
Safety canisters, electrically conducting
Supplier: S.C.A.T.
Avantor® ACE® C18-AR, colonne HPLC, 3 µm
Supplier: Avantor
Safety waste caps and ventilation kit, SymLine®
Supplier: S.C.A.T.
Aluminium blocks, Reacti-Block™
Supplier: Thermo Fisher Scientific