Bench top concentrator, 5301
Product Details & Documents
This concentrator is an easily upgradeable basic device which enables efficient and gentle vacuum concentration of samples. Optimal and rapid evaporation of your samples is made possible by innovative powerful heating technology.
- Chemical-resistant stainless steel chamber
- Temperature preselection: room temperature, 30, 45 and 60 °C
- Variable tube connection for external vacuum pump or central vacuum system
- Basic device easily upgradeable to the complete system
The emission condenser that is supplied with the concentrator purifies the exhaust air by up to 85% as a protection against unpleasant vapors and an automatic condensate drain prolongs the service life of all components that come into contact with vapors.
Delivery information: Basic device is supplied with 48×1,5/2,0 ml fixed-angle rotor. For upgrading the basic device to a complete system, a diaphragm pump with inner PTFE coating, an emission condenser and two rails for connecting concentrator and pump need to be ordered separately.
Specifiche
- Descrizione:Concentrator 5301, basic device, UK-plug
- Seq:01
- Cat. No.:EPPE5301000.067
- Peso:17 kg
- Velocità massima:1400 min⁻¹
- Temperatura:Ambient/30/45/60 °C
- Modello:5301 (basic)
- Vuoto max.:20 mbar
- Capacità max.:48×1,5/2,0 ml
- Potenza max. assorbita:150 W
- L×P×A:320×370×230 mm
- Rumorosità:≤60 dB(A)
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Distanziale per funzionamento sandwich dei rotori ad angolo fisso, per rotori F-45-72-8 e F-45-48-11Distanziale per funzionamento sandwich dei rotori ad angolo fisso, per rotori F-45-72-8 e F-45-48-11700-1384Log in per disponibilità1 * 1 ST - SFr. 36.20
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



