Orders LinkContactUsLinkComponent
Trolleys, Variocar® 60 basic
Trolleys, Variocar® 60 basic
Trolleys, Variocar® 60 basic
Trolleys, Variocar® 60 basic
Multipurpose trolleys with chromium plated frame and shelves.
  • White shelves and drawers with designed drawer handles in different colours
  • Castors of 75 mm Ø each
  • Total loading capacity up to 50 kg
Product Family Options
  • Descrizione
    L×P×A
  • Trolley, with one drawer
    600×450×730 mm
    1 * 1 ST
    1 * 1 ST - SFr. 1’260.00
    SFr. 1’260.00
    1 * 1 ST
       SFr. 1’260.00
  • Trolley, with three drawers
    600×450×730 mm
    1 * 1 ST
    1 * 1 ST - SFr. 1’850.00
    SFr. 1’850.00
    1 * 1 ST
       SFr. 1’850.00
  • Trolley, with four drawers
    600×450×730 mm
    1 * 1 ST
    1 * 1 ST - SFr. 2’210.00
    SFr. 2’210.00
    1 * 1 ST
       SFr. 2’210.00