Septa 8 mm for screw neck vials, 8-425
These septa are designed to be used with 8 mm screw thread caps.
Frequently Bought Together
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR®, Screw Neck Vials ND8
: VWR Collection
VWR®, Screw Caps for Short Thread Vials ND9
: VWR Collection
Acqua, HiPerSolv CHROMANORM® per HPLC
: VWR Chemicals
VWR®, Short Thread Vials ND9
: VWR Collection
VWR®, Screw Neck Vials ND24
: VWR Collection
VWR®, Snap Cap Vials
: VWR Collection
Acetone ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS per analisi
: VWR Chemicals
VWR®, Crimp Neck Vials ND11
: VWR Collection
VWR®, Screw Neck Vials ND13
: VWR Collection