Fisher Container™ Autoclavable Precision Clean® Breather Bags
Catalog # FISH90726P
Anbieter: FISHER CONTAINER
Technische Daten
- Descrizione:Autoclavable Precision Clean® Breather Bags
- L×L:210×310 mm
- Conf:500
Technische Daten
Über diesen Artikel
A combination of Tyvek™ and clear, ultraclean film structure aids in the prevention of interior particle contamination. Designed for products to be used in cleanroom environments that also require sterilization.
- Faster cycle times
- Molecular bonded seals
- Pouch suitable for sterilizaiton by steam, EtO, gamma radiation or hydrogen peroxide
- Tyvek™ 1059B one side and one side 3Mil PCHD
Certifications: Certified to NASA™ clean standards by Fisher to meet Level 100 KSC-C-123