Orders LinkContactUsLinkComponent
General purpose thermometers
Supplier:   Ludwig Schneider
General purpose thermometers
Supplier:  Ludwig Schneider
Certificates
General purpose thermometers
General purpose thermometers
11 Options Available Below
Product Details & Documents
These stem type thermometers with red filing are suitable for general purpose applications.
  • Permanent graduation and lettering
  • With loop
  • Capillary diameter: 6 ±1 mm
Product Family Options
Product Information
  • Descrizione
    Range di misura
    Divisione
    Lunghezza
    Riempimento
    Profondità d'immersione
    Ø
    Availability
    Price
  • Stem type, round, with white back, capillary form
    −35...+50 °C
    1,0 °C
    300 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 9.90
    SFr. 9.90
    1 * 1 ST
       SFr. 9.90
  • Stem type, round, with white back, capillary form
    −35...+50 °C
    1,0 °C
    300 mm
    Red
    76 mm
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 11.20
    SFr. 11.20
    1 * 1 ST
       SFr. 11.20
  • Stem type, round, with white back, capillary form
    −20...+110 °C
    1,0 °C
    300 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 9.90
    SFr. 9.90
    1 * 1 ST
       SFr. 9.90
  • Stem type, round, with white back, capillary form
    −20...+110 °C
    1,0 °C
    300 mm
    Red
    76 mm
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 11.20
    SFr. 11.20
    1 * 1 ST
       SFr. 11.20
  • Stem type, round, with white back, capillary form
    −20...+150 °C
    1,0 °C
    300 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 10.90
    SFr. 10.90
    1 * 1 ST
       SFr. 10.90
  • Stem type, round, with white back, capillary form
    −20...+150 °C
    1,0 °C
    300 mm
    Red
    76 mm
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 12.10
    SFr. 12.10
    1 * 1 ST
       SFr. 12.10
  • Stem type, round, with white back, capillary form
    −10...+60 °C
    1,0 °C
    200 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 9.70
    SFr. 9.70
    1 * 1 ST
       SFr. 9.70
  • Stem type, round, with white back, capillary form
    −10...+60 °C
    1,0 °C
    300 mm
    Red
    76 mm
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 10.90
    SFr. 10.90
    1 * 1 ST
       SFr. 10.90
  • Stem type, round, with white back, capillary form
    −10...+200 °C
    1,0 °C
    300 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 11.90
    SFr. 11.90
    1 * 1 ST
       SFr. 11.90
  • Stem type, round, with white back, capillary form
    −10...+200 °C
    1,0 °C
    300 mm
    Red
    76 mm
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 14.20
    SFr. 14.20
    1 * 1 ST
       SFr. 14.20
  • Stem type, round, with white back, capillary form
    −10...+250 °C
    1,0 °C
    300 mm
    Red
    Total
    6 ±1 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 17.50
    SFr. 17.50
    1 * 1 ST
       SFr. 17.50