Prescription sieves
Supplier: LINKER INDUSTRIE TECHN
Certificates
About this item
These prescription sieves are made from aluminium and stainless steel.
Made up of lid, 2 sieve rings and a collection pan.
Frequently Bought Together
These prescription sieves are made from aluminium and stainless steel.
Made up of lid, 2 sieve rings and a collection pan.