Heating tapes, IT-S20
Informazioni su questo articolo
Isopad IT-S20 is a glass silk, preterminated, flexible heating tape with a copper-tinned outer braiding serving as a protective earth braid. The temperature depends on the power and application. This enables maximum power to be installed even in the small surfaces. Provided with class I electrical protection.
- Maximum operating temperature: 450 °C
- Width×Thickness: 12×7 mm (±10%)
- Maximum power per meter 220 W/m
It is non hazardous and used in ordinary areas. It is used for temperature modification of pipes and structures in industries and laboratories.
6 varianti
Product Details & Documents
Isopad IT-S20 is a glass silk, preterminated, flexible heating tape with a copper-tinned outer braiding serving as a protective earth braid. The temperature depends on the power and application. This enables maximum power to be installed even in the small surfaces. Provided with class I electrical protection.
- Maximum operating temperature: 450 °C
- Width×Thickness: 12×7 mm (±10%)
- Maximum power per meter 220 W/m
It is non hazardous and used in ordinary areas. It is used for temperature modification of pipes and structures in industries and laboratories.
Product Family Options
Informazioni sul prodotto
- DisponibilitàPrice
- 454-3011Heating tape IT-S20, 1 m length, 220 W
- ISOP704368-000Heating tape IT-S20, 1,5 m length, 330 W
- ISOP153620-000Heating tape IT-S20, 2 m length, 440 W
- ISOP443830-000Heating tape IT-S20, 3 m length, 650 W
- ISOP054874-000Heating tape IT-S20, 5 m length, 1090 W
- ISOP261076-000Heating tape IT-S20, 10 m length, 2180 W