Parasitology prepared slides
Catalog # LIED74900
Supplier: LIEDER JOHANNES
Specifications
- Descrizione:Set of 25 slides
- Conf:1
Specifications
About this item
Blood smear (t.s) tranverse section (w.m) whole mount (l.s) longitudinal section.
- Sleeping disease (sm)
- Malaria in rodents (sm) with vegetative forms and schizogony stages
- Section of muscle showing the parasites in Miescher’s tubes
- Honey bee dysentery (t.s) of diseased bee intestine
- Causes coocidiosis in rabbit liver (t.s) shows parasites in all stages
- Beef liver fluke (w.m) of adult flat mount and carefully stained
- Fasciola hepatica, ova (w.m)
- Taenia spec. or Moniezia spec., tapeworm, scolex (head) (w.m)
- Dog tapeworm, mature proglottids (w.m).
- Tapeworm, proglottids in different stages (t.s)
- Dwarf tapeworm, proglottids (w.m)
- Cyst wall and scolices sec.
- Roundworm of human, adult female (t.s) in region of gonads
- Roundworm ova from faeces (w.m)
- Pin worm adult specimen (w.m)
- Muscle with encysted larvae (l.s)
- Tick adult (w.m) carrier of relapsing fever and borreliosis
- Chicken mite (w.m) adult specimen
- Parasitic mite of honey bee (w.m)
- Section of diseased skin with parasites
- Anopheles, malaria mosquito, head and mouth parts of female (w.m)
- Common mosquito, head and mouth parts of female (w.m)
- Bed bug (w.m) of male or female
- Human louse (w.m) of male or female
- Dog flea, adult male or female (w.m)
Ordering information: Includes 25 microscope slides with accompanying brochure.