Mixing measuring cylinders, tall form, class B, glass
Catalog # HACH2088640
Supplier: Hach
Specifications
- Capacità:25 ml
- Seq:01
- Divisione:0,5 ml
- Conf:1
- Tolleranza:0,2 ± ml
Specifications
About this item
Glass cylinder, Class B.
- Calibrated 'to contain' (TC)
- Molded hexagonal base
- Glass stopper
- Graduations in durable white enamel