Orders LinkContactUsLinkComponent
Cloning cylinders
Cloning cylinders
Nome del fornitore:   Chemicon
Cloning cylinders
Nome del fornitore:  Chemicon
Certificates
Cloning cylinders
2 varianti
Product Details & Documents
Borosilicate glass 3.3.
  • Sterile
  • Ready to use
  • Can be stored at room temperature
Individual colonies of transfected cells can be isolated and picked from a plate containing many clones. These sterile cloning cylinders are greased on one end to allow the cylinder to seal to the plate. Cells are then dissociated within the cylinder, resuspended, and transferred to a new vessel as a homogeneous population of cells derived from a single colony.
Product Family Options
Informazioni sul prodotto
  • Altezza
    Ø est
    Disponibilità
    Price
  • 8 mm
    8 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 15 ST
    1 * 15 ST - SFr. 220.00
    SFr. 220.00
    1 * 15 ST
       SFr. 220.00
  • 10 mm
    10 mm
    Log in per disponibilità
    Log in per disponibilità
    1 * 10 ST
    1 * 10 ST - SFr. 217.00
    SFr. 217.00
    1 * 10 ST
       SFr. 217.00