Orders LinkContactUsLinkComponent
OX™ Cover Spectacles
OX™ Cover Spectacles
Nome del fornitore:   3M
OX™ Cover Spectacles
Nome del fornitore:  3M
Certificates
OX™ Cover Spectacles
OX™ Cover Spectacles
1 varianti
Product Details & Documents
The 3M™ OX cover glasses (OTG) variants offer unique distinctive features: The OX2000 comes with adjustable lock-in temples (5 positions) and rotating tips (excellent grip in all positions) and the OX3000 is suitable for use with earmuffs; with low profile adjustable temples and non-slip pliable temple tips.
  • Optical Class 1 lens suitable for prolonged use
  • Protects against high speed particles at low energy (F) at extreme temperature conditions –5 and +55 °C (T)
  • Offers excellent protection against UV radiation
  • Designed to be worn over prescription eyewear
Can be used in a wide range of applications including construction, engineering, general assembly, light duty maintenance and repair and woodworking.
Certifications: EN 166:2001, EN 170:2002.
Product Family Options
Informazioni sul prodotto
  • Tipo
    Colore
    Materiale lente
    Disponibilità
    Price
  • OX 3000: For use with earmuffs; anti-UV (2C-1,2), anti-scratch
    blu
    Clear PC
    Discontinued
  • OX 3000: For use with earmuffs; anti UV (2C-1,2), anti-scratch
    blu
    Clear PC
    Log in per disponibilità
    Log in per disponibilità
    1 * 1 ST
    1 * 1 ST - SFr. 13.50
    SFr. 13.50
    1 * 1 ST
       SFr. 13.50