Cleanroom wipes, C-Wipes, C6
Catalog # EPTA7-C6-99L-00
Supplier: ESSENTRA POROUS TECHNOLOGIES
Specifications
- Confezione:500/bag
- L×L:230×230 mm
- Conf:500
Specifications
About this item
This lightweight, low linting tissue is made from a specialty blend of hemp and cellulose fibers. It is ideal for general purpose cleaning applications.
- Effective in the cleanroom because of their super absorbency, extremely low particle generation, and ultra purity
- Absorb four times their own weight
- Soft wipes will not scratch surfaces when used either wet or dry
- Low in soluble extractables and metallic ions
Colour: White
Packaging: Converted, cleaned, and double-bag packaged in a cleanroom environment.