Thermo Scientific™ Locator™ Plus Rack and Box Systems
Supplier: Thermo Fisher Scientific
Certificates
About this item
Locator™ cryogenic rack and box systems allow quick, efficient sample retrieval with maximum storage capacity (up to 6000 samples with Locator™ Plus) combined with minimum liquid nitrogen consumption.
- Compact design minimises space requirements while maximising storage capacity
- System is suited for both manual and computerised inventory record-keeping methods
- Super-insulated vessels have special, narrow-mouth design that conserves liquid nitrogen by minimising evaporation
- Outstanding temperature uniformity: Samples are stored below –180 °C, even when less than 5 cm of liquid nitrogen remains in the vessel
- Ultrasonic level monitor safeguards irreplaceable samples with minimal liquid nitrogen evaporation and conduction
- Secure locking clasp prevents unauthorised entry
- Audible alarm that sounds when nitrogen level falls below safe range; dry remote alarm contact for remote monitoring
Locator™ cryogenic rack and box systems feature a unique hanging rack and gridded box design, advanced vacuum insulation, unmatched temperature uniformity and an innovative liquid nitrogen monitor.
Locator™ Plus systems are available with or without ultrasonic level monitor.
Note: Stated vial capacity assumes 1,2/2 ml Thermo Scientific™ Nalgene™ cryogenic vials.
Ordering information: Requires Thermo Scientific™ Nalgene™ cryogenic boxes (order separately).
Delivery information: Locator™ systems include indexed stainless steel racks that fit into corresponding locations in the vessel. Each rack has individual storage compartments that hold one Thermo Scientific™ Nalgene™ cryogenic storage box.
Locator™ Plus systems include indexed stainless-steel racks designed for use with 5 cm Nalgene™ cryogenic boxes (larger racks available for storage of 5,0 ml ampoules in Locator Jr. Plus and Locator 4 Plus). Available with or without liquid nitrogen level monitor.
Locator™ Plus systems include indexed stainless-steel racks designed for use with 5 cm Nalgene™ cryogenic boxes (larger racks available for storage of 5,0 ml ampoules in Locator Jr. Plus and Locator 4 Plus). Available with or without liquid nitrogen level monitor.
Specifications
- Catalog No:
- BARNCY509111
- 478-0167
- BARNCY509109
- BARNCY509110
- 478-0265
- 478-0807
- BARNCY509107
- 478-0267
- BARNCY509108
- 478-0268
- 478-0266
- BARNCY509105
- BARNCY509106
- Modello:
- Locator 8 Plus
- Locator Jr.
- Locator 6 Plus
- Locator 8
- Locator Jr. Plus
- Locator 4
- Locator 4
- Locator 8 Plus
- Locator 4 Plus
- Locator 6 Plus
- Locator 4 Plus
- Locator Jr.
- Locator Jr. Plus
- Capacità:
- 121 L
- 60 L
- 184 L
- 111 L
- 71 L
- 111 L
- 111 L
- 121 L
- 121 L
- 184 L
- 121 L
- 60 L
- 71 L
- Autonomia statica:
- 201
- 70
- 185
- 185
- 83
- 112
- 112
- 201
- 122
- 185
- 122
- 70
- 83
- Evaporazione giornaliera:
- 0,60 L
- 0,85 L
- 0,99 L
- 0,60 L
- 0,85 L
- 0,99 L
- 0,99 L
- 0,60 L
- 0,99 L
- 0,99 L
- 0,99 L
- 0,85 L
- 0,85 L
- Ø del collo:
- 152 mm
- 215 mm
- 215 mm
- 152 mm
- 215 mm
- 215 mm
- 215 mm
- 152 mm
- 215 mm
- 215 mm
- 215 mm
- 215 mm
- 215 mm
- Ø×H:
- 558×953 mm
- 558×622 mm
- 660×953 mm
- 558×953 mm
- 558×683 mm
- 558×902 mm
- 558×902 mm
- 558×953 mm
- 558×953 mm
- 660×953 mm
- 558×953 mm
- 558×622 mm
- 558×683 mm