Multipurpose basins, with handles
Catalog # SEMA1752
Supplier: SEMADENI
Specifications
- Capacità:11,0 l
- Colore:grigio
- L×P×A:390×300×150 mm
- Conf:1
Specifications
About this item
PP, granite.
- Rectangular, with two side handles
- Stackable and resistant in hot water
- Suitable for food
Multipurpose basins for a wide range of applications.