ContactUsLinkComponent
Paper Bags
  KING626251/001
undefined
Paper Bags
  KING626251/001
  • Descrizione:
    Paper bag with gusset , white
  • Materiale:
    Paper
  • Seq:
    01
  • L×P×A:
    250×380×100 mm
  • Conf:
    500
Packaging paper bag with gusset.
 : EN 868-1 and 868-4