Pipette tips
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
- Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
- Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
- With graduations to visually check volume
- Suitable for all major pipettor brands
These tips are manufactured from medical grade PP in modern production facilities with stringent quality controls at every stage.
- Tips are lot tested and certified for absence of detectable human DNA and detectable endotoxins (pyrogens)
- Tips are tested and certified for absence of heavy metals, detectable DNase and RNase
- With graduations to visually check volume
- Suitable for all major pipettor brands
Product Family Options
- Stile puntaVolumeSteriliLunghezzaCodice coloreConfezione
- Micro, ultrafine0,1 - 10 µl-32 mmnaturale96 tips/rack; 12 racks/case1 * 1.152 STSFr. 209.00
- Micro, ultrafine0,1 - 10 µl+32 mmnaturale96 tips/rack; 12 racks/case1 * 1.152 STSFr. 236.00
- Micro, ultrafine0,1 - 10 µl-32 mmnaturaleBulk, 1000 tips/bag1 * 1.000 STSFr. 64.30
- Bevelled, non-graduated1 - 200 µl-50 mmgiallo96 tips/rack; 10 racks/case1 * 960 STSFr. 172.00
- Bevelled, non-graduated1 - 200 µl+50 mmgiallo96 tips/rack; 10 racks/case1 * 960 STSFr. 198.00
- Bevelled, non-graduated1 - 200 µl-50 mmgialloBulk, 1000 tips/bag1 * 1.000 STSFr. 64.30
- Bevelled, graduated1 - 200 µl-50 mmnaturaleBulk, 1000 tips/bag1 * 1.000 STSFr. 64.30
- Bevelled, graduated1 - 200 µl+50 mmnaturale96 tips/rack; 10 racks/case1 * 960 STSFr. 210.00
- Bevelled, non-graduated200 - 1000 µl-76 mmblu96 tips/rack; 6 racks/case1 * 576 STSFr. 151.00
- Bevelled, non-graduated200 - 1000 µl-76 mmbluBulk, 1000 tips/bag1 * 1.000 STSFr. 108.00
- Bevelled, non-graduated200 - 1000 µl+76 mmblu96 tips/rack; 6 racks/case1 * 576 STSFr. 171.00