Cleanroom gloves, latex, hand-specific, Kimtech™ G3
Supplier: KIMBERLY CLARK
Certificates
About this item
Hand-specific gloves with rolled, beaded cuffs for easy donning and better seal on garment.
- Non sterile, powder-free, textured finish
- Washed repeatedly in ultrapure deionised water to ensure consistent control of particles and extractables
- Exceptional wet/dry grip without compromising tactile sensitivity
Suitable for use in class 1, ISO 3, M1.5 cleanrooms.
EN 374, Cat. III; AQL 1.5
Certifications: PPE Cat III (EU) 2016/425; EN 374-1 Type C (K), EN 374-5 (VIRUS); AQL 1.5
Packaging: 50 pairs per bag.
Frequently Bought Together