Cleanroom wipes, pre-saturated, sterile, PROSAT® Sterile™ Low Endotoxin
Supplier: CONTEC
Certificates
About this item
These low endotoxin wipes are made from 100% knitted polyester and pre-saturated with a blend of 70% USP-Grade IPA and 30% water for injection (WFI).
- Laser-sealed edges
- Good abrasion and chemical resistance
- Pouch can be easily resealed throughout use maintaining saturation levels
- Each batch of wipes has a guaranteed endotoxin level of less than 1 EU/wipe
- Validated sterile via gamma irradiation to a sterility assurance level (SAL) of 10⁻⁶
Suitable for use in the most critical pharmaceutical cleanrooms.
Suitable for use in Grade A, ISO 3 Class 100, ISO 5, M3.5 cleanrooms.
Colour: White
Caution: Data accurate at time of publishing, latest test results and test methods can be found on supplier’s website.
Related Information
Certificates
Search by Lot NumberCoC_CONTPSWE0001-BPR_1031086 CoC_CONTPSWE0001-BPR_890980-0-0-1 CoC_CONTPSWE0002-BPR_853967-0-0-1 CoC_CONTPSWE0003-BPR_853968-0-0-1 CoC_CONTPSWE0003-BPR_868665-0-0-1
Mehr
CONTPSWE0001-BPR_TDS_EN
Technisches Datenblatt
Endotoxin control – consumable products that can help Endotoxins and Endotoxin control It’s just a Mop? Choice of Mopping Systems
Low Endotoxin Products for Critical Environments Contec
Produktliteratur
Frequently Bought Together