ContactUsLinkComponent
Octenisan®, 2-in-1 Antibacterial Body Wash and Shampoo
Octenisan®, 2-in-1 Antibacterial Body Wash and Shampoo
Codice catalogo SCUE121501
Nome del fornitore:  SCHÜLKE & MAYR GMBH
undefined
Octenisan®, 2-in-1 Antibacterial Body Wash and Shampoo
Codice catalogo SCUE121501
Nome del fornitore:  SCHÜLKE & MAYR GMBH
Specifiche
  • Descrizione:
    Octenisan® antibacterial shampoo and body wash, 150 ml bottle
  • Seq:
    01
  • Conf:
    30
Specifiche
Informazioni su questo articolo
Mild and gentle antimicrobial shampoo and body wash. Suitable for all skin types, also for all over body washing to combat MRSA. Particularly suitable for hospital use in intensive care and infectious diseases units.
  • Contains specially selected skin care ingredients, gentle surfactants and octenidine dihydrochloride
  • Skin-neutral pH
  • Free of colourant and perfume
Caution: Use biocides safely. Always read the label and product information before use.
*I prodotti in questa pagina non rientrano nell'ambito della Direttiva UE 2019/904