ContactUsLinkComponent
Hand cleansing pastes, IVRAXO® SOFT G
Hand cleansing pastes, IVRAXO® SOFT G
Catalog # FAWE11957015
Supplier:  FAWECO HAUTSCHUTZ
undefined
Hand cleansing pastes, IVRAXO® SOFT G
Catalog # FAWE11957015
Supplier:  FAWECO HAUTSCHUTZ
Specifications
  • Descrizione:
    Hand cleansing paste, IVRAXO® SOFT G, in 250 ml bottle
  • Conf:
    250 mL
Specifications
About this item
Hand cleansing paste contains plastic scrubbers and is suitable for heavy dirt like greases, oils, metallic dust, graphite or soot.
  • Thorough but gentle cleansing
  • Adapted to the natural pH-value of human skin
  • Solvent- and soap-free
  • Perfumed or fragrance-free
  • Re-moisturising
Caution: *I prodotti in questa pagina non rientrano nell'ambito della Direttiva UE 2019/904