Cleaner for instruments, Helizyme
Codice catalogo BRAU18557
Nome del fornitore: B. Braun
Specifiche
- Descrizione:Helizyme, enzymatic manual cleaner
- Conf:1 L
Specifiche
Informazioni su questo articolo
Helizyme is a pH neutral enzymatic cleaner with the combination of enzymes and surfactants, used for manual cleaning of resuable, invasive and non-invasive medical devices (surgical instruments, flexible and rigid endoscopes).
- Excellent material compatibility
- Suitable for ultrasonic cleaning
- Clinically proven biofilm removal