Disinfectants for cleanrooms, ClearKlens Bi-Spore
141-1449
: Diversey
- Descrizione:Disinfectant, ClearKlens Bi-Spore VH26
- Seq:02
- Confezione:25 x (100 ml base concentrate + 100 ml activator)
- Conf:25 SET
ClearKlens Bi-Spore is a rapid action, stable, two part chlorine dioxide generating system, developed to disinfect pharmacuetical industry sterile production facilities. It passes the European sporicidal test (CEN 13704) in 5 minutes, well within realistic cleanroom surface drying times, 5 to 8 minutes after surface contact.
- 0,2 µm filtered, aseptically filled and double-bagged
- Easy to use two-part system- base and activator
- Compatible with all surfaces typically found in cleanrooms
- Supplied with Certificate of Analysis and sterility where applicable
- Complimentary rotational partner to ClearKlens Tego® 2000, ClearKlens Tego® 2001 and ClearKlens Cleansinald
: Bactericide EN 1276; Fungicide EN 1650; Bactericide and Fungicide EN 13697; Sporicide EN 13704; Virucide EN 14476.
: Use biocides safely. Always read the label and product information before use.