Wipes, pre-saturated
152×216 mm, 100/canister
Supplier: ESSENTRA POROUS TECHNOLOGIES
Jump to:
Product Details & Documents
These polyester/cellulose wipes are pre-saturated with 96% IPA and 4% DIW. They are ideal for general and manufacturing applications, as well as for industrial cleaning.
- Perforated roll pre-wetted with alcohol
- Canister with spring loaded lid prevents dying out and facilities dispensing
- Alcohol saturates and evaporates quickly
- Made in the USA
: *I prodotti in questa pagina non rientrano nell'ambito della Direttiva UE 2019/904
Specifications
- Confezione:100/canister
- L×L:152×216 mm
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.