Hybridisation bottles
Hybridisation bottles, GL 45 screw cap with silicone sealing, 38×150 mm, Hybridisation ovens type Hybaid, Bellco, Labline, GFL, Heraeus
Codice catalogo WITG2.446.115
Nome del fornitore: witeg Labortechnik
Vai a
Product Details & Documents
Borosilicate glass 3.3, with GL 45 screw cap and silicone sealing.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Specifiche
- Ø×L:38×150 mm
- Per:Hybridisation ovens type Hybaid, Bellco, Labline, GFL, Heraeus
- Descrizione:Hybridisation bottles, GL 45 screw cap with silicone sealing