Detergent, liquid concentrate, Witonex-B-26
Codice catalogo WITG5.220.010
Nome del fornitore: witeg Labortechnik
Specifiche
- Descrizione:Witonex-B-26, liquid detergent, for rinsing washing machines
- Environmentally Preferable:
- Conf:10 kg
Specifiche
Informazioni su questo articolo
Liquid concentrate specially formulated to be low foaming for use with laboratory dishwashers.
- For routine cleaning of objects and apparatus in medical, chemical and technical laboratories
- Suitable for objects made of porcelain, glass, plastic, rubber and metals