Spatulas, Trulla type
Trulla spatulas, pattern type, 175 mm
Codice catalogo NICK9470
Nome del fornitore: NICKEL ELECTRO
Vai a
Product Details & Documents
Stainless steel, PTFE XYLAN® coated. These spatulas feature a PTFE XYLAN® coating which provides a non stick surface to which very few solid or liquid substances will permanently adhere.
- Stainless steel base: Spatula can withstand extreme temperatures and chemicals
- PTFE coating provides chemical inertness, purity, low permeability, heat resistance, toughness, and non stick qualities
- Excellent non wetting properties make cleaning very easy
- Spatula can undergo steam or chemical sterilisation
Ideal for biology, microbiology and pharmaceutical applications.
Specifiche
- Descrizione:Trulla spatulas, pattern type
- Lunghezza:175 mm