Container with Wooden Pallet
N° de catalogue SEMA2082
Fournisseur: SEMADENI
Spécifications
- Capacità:1000 l
- L×P×A:1200×1000×1170 mm
- Conf:1
Spécifications
À propos de cet article
HDPE, natural
- UV stabilised for storage outside
- Robust tubular steel frame provides additional protection during transport
- Sealable in- and outlet
Space-saving and transportable container for fluids and chemicals.