Clamps, three pronged
Clamps, three pronged, Alluminio, 0 - 100 mm, 140 mm
Codice catalogo BOCH5565
Nome del fornitore: BOCHEM
Vai a
Product Details & Documents
Three pronged retort clamp made of aluminium.
- Fingers of the clamps are covered with a layer of cork to protect clamped items
Specifiche
- Larghezza di serraggio:0 - 100 mm
- Descrizione:Clamps, three pronged
- Lunghezza:140 mm
- Materiale:Alluminio