Waste Bins
Informazioni su questo articolo
Made of HDPE.
- Stable and are stackable
- Odourless and hermetic seal for the transport and disposal
- Adhesive sealing and triple lid seal
Round containers are suitable for safe and clean disposal of clinical, bacteriological or chemical and hazardous waste.
Certifications: UN 1H2/Y11/S (30 litre bins), max. loading 11 kg
UN 1H2/Y22/S (60 litre bins), max. loading 22 kg
Informations de commande: Lids must be ordered separately.
2 varianti
Product Details & Documents
Made of HDPE.
- Stable and are stackable
- Odourless and hermetic seal for the transport and disposal
- Adhesive sealing and triple lid seal
Round containers are suitable for safe and clean disposal of clinical, bacteriological or chemical and hazardous waste.
Certifications: UN 1H2/Y11/S (30 litre bins), max. loading 11 kg
UN 1H2/Y22/S (60 litre bins), max. loading 22 kg
Informations de commande: Lids must be ordered separately.
Product Family Options
Informazioni sul prodotto
- ColoreMaterialeCapacitàØ×ADisponibilitàPrice
- SEMA2092Waste disposal bin, without lidWaste disposal bin, without lidantraciteHDPE30 L400×350 mmLog in per disponibilità1 * 1 STSFr. 28.80
- SEMA2093Waste disposal bin, without lidWaste disposal bin, without lidantraciteHDPE60 L400×700 mmLog in per disponibilità1 * 1 STSFr. 37.30
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Lid for waste bin, yellow
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Lid for waste bin, anthracite
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Lid for waste bin, yellow
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Lid for waste bin, red
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.