Silicone sponge stoppers, neoLab®
Jump to:
About this item
These silicone sponge stoppers are designed for closing round containers or bottle necks.
- Chemical and thermal properties of silicone
- Light as a feather, autoclavable, odourless and non-toxic
- Resistant to temperatures between −60 and +200 °C
0 Options Available Below
Product Details & Documents
These silicone sponge stoppers are designed for closing round containers or bottle necks.
- Chemical and thermal properties of silicone
- Light as a feather, autoclavable, odourless and non-toxic
- Resistant to temperatures between −60 and +200 °C